Learn More
NUSAP1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12024-20UL
 View more versions of this product
Product Code. 18074805
  • £135.00 / 20µL
Description
The NUSAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NUSAP1. This antibody recognizes human antigen. The NUSAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
NUSAP1
1000 μg/mL
ANKT, BM037, FLJ13421, LNP, NuSAP, NUSAP1, PRO0310, PRO0310p1, Q0310, SAPL
20 μL
51203
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa