Learn More
NUSAP1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-12024-100UL
Description
The NUSAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NUSAP1. This antibody recognizes human antigen. The NUSAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NUSAP1 | |
| 1000 μg/mL | |
| ANKT, BM037, FLJ13421, LNP, NuSAP, NUSAP1, PRO0310, PRO0310p1, Q0310, SAPL | |
| 100 μL | |
| 51203 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |