missing translation for 'onlineSavingsMsg'
Learn More

NUSAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-68989-25ul

 View more versions of this product

Product Code. 18649556

  • £328.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NUSAP1 Polyclonal antibody specifically detects NUSAP1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUSAP1
Polyclonal
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
ANKTNUSAP, BM037, FLJ13421, LNP, nucleolar and spindle associated protein 1, nucleolar and spindle-associated protein 1, nucleolar protein ANKT, NuSAP, NuSAP1, PRO0310p1, Q0310, SAPL
This antibody was developed against a recombinant protein corresponding to amino acids: THKLKTITGNSAAVITPFKLTTEATQTPVSNKKPVFDLKASLSRPLNYEPHKGKLKPWGQSKENNYLNQHVNRINFYKKTYKQPHLQ
25 μL
Core ESC Like Genes, Stem Cell Markers
51203
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.