missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUSAP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06564MFV450
This item is not returnable.
View return policy
Description
NUSAP1 Polyclonal antibody specifically detects NUSAP1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NUSAP1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| ANKTNUSAP, BM037, FLJ13421, LNP, nucleolar and spindle associated protein 1, nucleolar and spindle-associated protein 1, nucleolar protein ANKT, NuSAP, NuSAP1, PRO0310p1, Q0310, SAPL | |
| Produced in rabbits immunized with E. coli-derived Human NUSAP1 fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51203 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction