missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUS1 Recombinant Rabbit Monoclonal Antibody (K01_1U84)
GREENER_CHOICE

Product Code. 30827893
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827893 100 μL
30828090 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827893 Supplier Invitrogen™ Supplier No. MA563705
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NUS1 Recombinant Monoclonal Antibody for Western Blot

With DHDDS, forms the dehydrodolichyl diphosphate synthase (DDS) complex, an essential component of the dolichol monophosphate (Dol-P) biosynthetic machinery. Adds multiple copies of isopentenyl pyrophosphate (IPP) to farnesyl pyrophosphate (FPP) to produce dehydrodolichyl diphosphate (Dedol-PP), a precursor of dolichol which is utilized as a sugar carrier in protein glycosylation in the endoplasmic reticulum (ER). Regulates the glycosylation and stability of nascent NPC2, thereby promoting trafficking of LDL-derived cholesterol. Acts as a specific receptor for the N-terminus of Nogo-B, a neural and cardiovascular regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUS1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1U84
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene NUS1
Gene Accession No. Q96E22, Q99LJ8
Gene Alias 1600027K07Rik; AU019165; AW538011; BC003223; C6orf68; Cis-prenyltransferase subunit NgBR; D10Ertd438e; dehydrodolichyl diphosphate syntase complex subunit NUS1; dehydrodolichyl diphosphate syntase complex subunit NUS1 pseudogene; dehydrodolichyl diphosphate synthase complex subunit NUS1; di-trans,poly-cis-decaprenylcistransferase; MGC:7199; NgBR; Nogo-B receptor; Nuclear undecaprenyl pyrophosphate synthase 1 homolog; nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae); Nus1; NUS1 dehydrodolichyl diphosphate synthase subunit; NUS1, dehydrodolichyl diphosphate synthase subunit; RGD1307879; TANGO14; transport and golgi organization 14 homolog
Gene Symbols NUS1
Host Species Rabbit
Immunogen A synthetic peptide of human Nogo B receptor.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116150, 294400, 52014
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.