missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nurr1/NGFI-B beta/NR4A2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40415PCP
This item is not returnable.
View return policy
Description
Nurr1/NGFI-B beta/NR4A2 Polyclonal antibody specifically detects Nurr1/NGFI-B beta/NR4A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Nurr1/NGFI-B beta/NR4A2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| human homolog of, nuclear receptor subfamily 4, group A, member 2, orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, T-cell nuclear receptor NOT, transcriptionally inducible nuclear receptor related 1, Transcriptionally-inducible nuclear receptor | |
| Synthetic peptide comprising residues 513-526 [TERHGLKEPKRVEE] of the human Nurr1 protein. Based on 100% sequence homology, this antibody is also expected to cross react with rat and mouse Nurr1. | |
| 0.1 mL | |
| Immune System Diseases, Immunology | |
| 4929 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction