missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nurim Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567247
This item is not returnable.
View return policy
Description
Immunogen sequence: HGLDQQDLRYL RAQLQRKLHL LSRP Highest antigen sequence identity to the following orthologs - mouse 96%, rat 92%.
Nurim is an inner nuclear membrane (INM) protein that is tightly associated with the inner nuclear membrane. It was first isolated in a visual screen for nuclear envelope-localizing proteins. In subcellular fractionation, nurim remained extremely tightly bound to nuclear fractions. Unlike the known nuclear envelope (NE) membrane proteins, it is neither associated with nuclear pores, nor targeted like lamin-associated membrane proteins suggesting its localization to NE by a distinct mechanism.
Specifications
| Nurim | |
| Polyclonal | |
| Unconjugated | |
| NRM | |
| 2610307M02Rik; AI429796; DAAP-285E11.8; Nrm; NRM29; Nuclear envelope membrane protein; nuclear rim protein; Nurim; nurim (nuclear envelope membrane protein); UNQ555/PRO1112 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IXM6 | |
| NRM | |
| Recombinant Human Nurim. Recombinant protein control fragment (Product #RP-105565). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction