missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16587-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for the export of mRNAs containing poly(A) tails from the nucleus into the cytoplasm. In case of infection by HIV-1, it may participate in the docking of viral Vpr at the nuclear envelope.Specifications
| NUPL2 | |
| Polyclonal | |
| Unconjugated | |
| NUPL2 | |
| CG1, hCG1, NLP 1, NLP_1, NLP1, Nucleoporin hCG1, nucleoporin like 2, Nucleoporin like protein 2, NUP42 homolog, NUPL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11097, 231042, 499974 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15504, Q8CIC2 | |
| Nupl2 | |
| NUPL2 Fusion Protein Ag8816 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction