Learn More
NUPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16587-1-AP-150UL
 View more versions of this product
Product Code. 16846244
  • £349.00 / 150µL
Description
Required for the export of mRNAs containing poly(A) tails from the nucleus into the cytoplasm. In case of infection by HIV-1, it may participate in the docking of viral Vpr at the nuclear envelope.
Specifications
Specifications
NUPL2
Polyclonal
Unconjugated
NUPL2
CG1, hCG1, NLP 1, NLP_1, NLP1, Nucleoporin hCG1, nucleoporin like 2, Nucleoporin like protein 2, NUP42 homolog, NUPL2
Rabbit
Antigen Affinity Chromatography
RUO
11097, 231042, 499974
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15504, Q8CIC2
Nupl2
NUPL2 Fusion Protein Ag8816
150 μL
Primary
Human, Mouse, Rat