Learn More
Invitrogen™ NUPL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588035
Description
Immunogen sequence: MAICQFFLQG RCRFGDRCWN EHPGARGAGG GRQQPQQQPS GNNRRGWNTT SQRYSNVIQP SSFSKSTPWG GSRDQEKPYF SSFDSGASTN RKEGFGLSEN PFASLSPDEQ KDEKKLLEGI VKDMEVWESS GQWMFSVYSP VKKKPNISGF TDISPEELRL EYHNFLTSNN LQSYLNSVQR LINQWRNRVN ELKSLNISTK; Positive Samples: HeLa, A-549, A-431, SGC-7901; Cellular Location: Cytoplasmic side, Nucleus, Nucleus membrane, Peripheral membrane protein, nuclear pore complex.
Required for the export of mRNAs containing poly(A) tails from the nucleus into the cytoplasm. [UniProt].
Specifications
| NUPL2 | |
| Polyclonal | |
| Unconjugated | |
| NUP42 | |
| AV116043; CG1; h H_RG271G13.9; H_RG271G13.9; hCG1; NLP_1; Nlp1; NLP-1; nucleoporin hCG1; nucleoporin like 2; Nucleoporin NUP42; Nucleoporin-42; nucleoporin-like protein 1; nucleoporin-like protein 2; NUP42; NUP42 homolog; Nupl2; RGD1560934 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15504 | |
| NUP42 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human NUPL2 (NP_0313681). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.