missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUPL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113478
This item is not returnable.
View return policy
Description
NUPL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the nucleoporin family that shares 87% sequence identity with rat nucleoporin p58. The protein is localized to the nuclear rim and is a component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| NUPL1 | |
| Polyclonal | |
| Unconjugated | |
| NUP58 | |
| 1700017F11Rik; 58 kDa nucleoporin; KIAA0410; mKIAA0410; nucleoporin 58; nucleoporin 58kDa; nucleoporin like 1; nucleoporin p45; nucleoporin p58; nucleoporin p58/p45; nucleoporin-like 1; nucleoporin-like protein 1; NUP45; NUP58; Nupl1; p58/p45; PRO2463 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 71844, 9818 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8R332, Q9BVL2 | |
| NUP58, Nupl1 | |
| Recombinant Human Nucleoporin p58/p45 protein (211-480AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction