Learn More
Invitrogen™ NuPAGE™ WedgeWell™ Tris-Acetate Welcome Pack and Mini Blot Modules, 3 to 8%
Description
The NuPAGE Tris-Acetate Welcome Pack and Mini Blot Modules provide all the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot modules for wet transfer.
The NuPAGE Tris-Acetate WedgeWell Welcome Pack and Mini Blot Modules provide all the necessary NuPAGE gels and buffers needed to begin using the Mini Gel Tank plus two Mini Blot modules for wet transfer.
Welcome Pack contents
- Mini Gel Tank (A25977)
- NuPAGE Tris-Acetate WedgeWell mini gels (2 boxes, 20 gels)
- NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
- NuPAGE LDS Sample Buffer, 4X (NP0007)
- NuPAGE Sample Reducing Agent, 10X (NP0009)
- HiMark Pre-stained Protein Standard (LC5699)
- 2 x Mini Blot Module (B1000)
About the Mini Gel Tank
The Mini Gel Tank is compatible with Novex gels, NuPAGE gels, and Bolt Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use.
About NuPAGE Tris-Acetate gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.
Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (Cat. No. NP0006) for traditional wet transfer using the Mini Blot Module (B1000).
About the Mini Blot Module
The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber or two blot modules total with the side-by-side layout. The Mini Blot Module is compatible all Invitrogen mini gels (blot size: 8.5 x 8 cm). The blot module accommodates one mini gel per wet transfer. The universal connection and molded gasket make the module easy to use, while the inner core of the module allows for use of less methanol-based transfer buffer per wet transfer (250 mL per gel). At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 60 minutes.
Convenient to use
NuPAGE Tris-Acetate WedgeWell-format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
Specifications
Specifications
| For Use With (Equipment) | XCell SureLock Mini-Cell |
| Gel Size | Mini |
| Product Line | NuPAGE |
| Gel Percentage | 3 to 8% |
| Gel Type | Tris-Acetate |
| No. of Wells | 12-well |
| Quantity | 1 Welcome Pk. kit |
| Separation Range | 40 to 500 kDa |
| Shipping Condition | Approved for shipment at room temperature and wet ice |
| Thickness (Metric) | 1.0 mm |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.