Learn More
Invitrogen™ NuPAGE™ Tris-Acetate Welcome Pack, 7%
The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents needed to begin using the Mini Gel Tank.
£656.00
Specifications
For Use With (Equipment) | Mini Gel Tank |
---|---|
Gel Percentage | 7% |
Gel Size | Mini |
GelThickness | 1.0 mm |
Gel Type | Tris-Acetate |
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
15824042
|
Invitrogen™
EA0355A |
10-well |
£656.00
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
15834042
|
Invitrogen™
EA0355B |
12-well |
£656.00
Each |
In Stock
Log in to see stock. |
|||||
15844042
|
Invitrogen™
EA0355C |
15-well |
£656.00
Each |
In Stock
Log in to see stock. |
|||||
Description
The NuPAGE Tris-Acetate Welcome Pack provides all of the necessary NuPAGE Tris-Acetate gels, buffers, and reagents you will need to begin using the Mini Gel Tank. The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading, and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.
The NuPAGE Tris-Acetate Welcome Pack contains:• Mini Gel Tank
• NuPAGE Tris-Acetate Mini gels (2 boxes, 20 gels)
• NuPAGE Tris-Acetate SDS Running Buffer (20X)
• NuPAGE LDS Sample Buffer (4X)
• NuPAGE Sample Reducing Agent (10X)
• HiMark™ Pre-stained Protein Standard
Mini Gel Tank
The Mini Gel Tank is compatible with all Novex, NuPAGE and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies being used.
NuPAGE Tris-Acetate Gels
NuPAGE Tris-Acetate protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate protein gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional laemmli system. Its unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine SDS-PAGE gels.
Transfer and detection
NuPAGE Tris-Acetate gels are compatible with the Mini Blot Module, Pierce Power Blotter, iBlot 2 Gel Transfer Device, iBind Western Device, and iBind Flex Western Device for optimal transfer and detection of proteins.
Specifications
Mini Gel Tank | |
Mini | |
Tris-Acetate | |
NuPAGE™ | |
Approved for shipment at room temperature and wet ice |
7% | |
1.0 mm | |
36 to 500 kDa | |
1 Welcome Pk. |