missing translation for 'onlineSavingsMsg'
Learn More

NUP98 Rabbit anti-Human, Mouse, Rat, Clone: S01-6F4, Novus Biologicals™

Product Code. 18376994 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376994 25 μg 25µL
18312302 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18376994 Supplier Novus Biologicals Supplier No. NBP31507025UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NUP98 Monoclonal antibody specifically detects NUP98 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP98
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S01-6F4
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96
Host Species Rabbit
Immunogen Recombinant protein of human NUP98 (Uniprot#: P52948)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4928
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.