missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NUP98 Antibody [Allophycocyanin], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-93327APC
This item is not returnable.
View return policy
Description
NUP98 Polyclonal antibody specifically detects NUP98 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NUP98 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| APC | |
| ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96 | |
| The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human nucleoporin 98kDa using the numbering given in entry NP_057404.2 (GeneID 4928). | |
| 0.1 mL | |
| Cancer, Mitotic Regulators | |
| 4928 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction