missing translation for 'onlineSavingsMsg'
Learn More

NUP98 Antibody (4G2), Novus Biologicals™

Product Code. 18327318 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327318 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327318 Supplier Novus Biologicals Supplier No. H00004928M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUP98 Monoclonal antibody specifically detects NUP98 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP98
Applications ELISA
Classification Monoclonal
Clone 4G2
Conjugate Unconjugated
Dilution ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12906
Gene Alias ADAR2, ADIR2, GLFG-repeat containing nucleoporin, nuclear pore complex protein Nup98-Nup96, nucleoporin 98kD, nucleoporin 98kDa, NUP196, NUP96, Nup98-Nup96
Host Species Mouse
Immunogen NUP98 (AAH12906.1, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKLYQTPLELKLKHSTVHVDELCPLIVPNLGVAVIHDYADWVKEASGDLPEAQIVKHWSLTWTLCEALWGHLKELDSQLNEPREYIQILERRRAFSRWLSCTATPQIEEE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4928
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.