missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP93 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | NUP93 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607802
|
Novus Biologicals
NBP2-94888-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651412
|
Novus Biologicals
NBP2-94888-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP93 Polyclonal antibody specifically detects NUP93 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NUP93 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9688 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| KIAA0095Nucleoporin Nup93,93 kDa nucleoporin, MGC21106, nuclear pore complex protein Nup93, nucleoporin 93kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 670-819 of human NUP93 (NP_055484.3). IAERYRAQGISANKFVDSTFYLLLDLITFFDEYHSGHIDRAFDIIERLKLVPLNQESVEERVAAFRNFSDEIRHNLSEVLLATMNILFTQFKRLKGTSPSSSSRPQRVIEDRDSQLRSQARTLITFAGMIPYRTSGDTNARLVQMEVLMN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title