missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP88 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
£161.00 - £341.00
Specifications
| Antigen | NUP88 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17999572
|
Invitrogen™
PA5145282 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17948772
|
Invitrogen™
PA5145281 |
200 μL |
£341.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). Nup88 (nucleoporin 88 kDa) is a 741 amino acid protein that localizes to the nucleus and functions as an essential component of the nuclear pore complex. Expressed ubiquitously, Nup88 is subject to phosphorylation by ATM or ATR and is upregulated in malignant neoplasms and precancerous dysplasias, suggesting a role in tumorigenesis. The gene encoding Nup88 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1, 200 genes.Specifications
| NUP88 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q8CEC0, Q99567 | |
| 19069, 4927 | |
| Recombinant Mouse NUP88 protein, Leu57-Gly296 (Accession #Q5QNU0). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 88 kDa nuclear pore complex protein; 88 kDa nucleoporin; karyoporin; nuclear pore complex protein 88; nuclear pore complex protein Nup88; nucleoporin 88; nucleoporin 88kDa; nucleoporin Nup84; Nucleoporin Nup88; Nup84; NUP88; Prei2; preimplantation protein 2 | |
| NUP88 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| NUP88 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title