missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP62CL Recombinant Rabbit Monoclonal Antibody (K01_2K66)
GREENER_CHOICE

Product Code. 30826381
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826381 100 μL
30826598 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826381 Supplier Invitrogen™ Supplier No. MA563961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NUP62CL Recombinant Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP62CL
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2K66
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene NUP62CL
Gene Accession No. Q9H1M0
Gene Alias nucleoporin 62 C-terminal like; nucleoporin 62kDa C-terminal like; nucleoporin-62 C-terminal-like protein; NUP62CL; NUP62L
Gene Symbols NUP62CL
Host Species Rabbit
Immunogen A synthetic peptide of human NUP62CL.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54830
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.