Learn More
Invitrogen™ NUP62 Monoclonal Antibody (RL1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1071
Description
MA1-071 detects nuclear pore-O-linked glycoprotein from rat, amphibian and yeast. MA1-071 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects up to eight different proteins from the nuclear pore complex (NPC) of approximately 210, 180, 145, 100, 63, 58, 54 and 45 kDa. Immunofluorescence staining of NPC O-linked glycoproteins with MA1-071 results in exclusive labeling of the NPC proteins on a wide variety of mammalian cells as well as S. cerevisiae and Xenopus. Labeling occurs exclusively at the NPC with most of the labeling at the cytoplasmic and nucleoplasmic margins. Microinjected MA1-071 inhibits both protein import and RNA export in Xenopus oocytes. The MA1-071 immunogen is pore complex-lamina fraction purified from rat liver nuclear envelopes.
SDS3 is a subunit of the histone deacetylase.
Specifications
| NUP62 | |
| Monoclonal | |
| Unconjugated | |
| Nup62 | |
| 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| 65274 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| RL1 | |
| PBS with 0.05% sodium azide | |
| P17955 | |
| Nup62 | |
| Pore complex-lamina fraction purified from rat liver nuclear envelopes. | |
| 100 μL | |
| Primary | |
| Rat, Amphibian, Yeast | |
| Antibody | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.