missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP62 Recombinant Rabbit Monoclonal Antibody (HL1226)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546961
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SDS3 is a subunit of the histone deacetylase.
Specifications
| NUP62 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P17955, P37198, Q63850 | |
| Nup62 | |
| Recombinant protein encompassing a sequence within the center region of human Nucleoporin p62. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1226 | |
| Unconjugated | |
| Nup62 | |
| 62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18226, 23636, 65274 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction