missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62 Antibody (RL31), Biotin, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
£441.00
Specifications
| Antigen | NUP62 |
|---|---|
| Clone | RL31 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
NUP62 Monoclonal antibody specifically detects NUP62 in Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| NUP62 | |
| Western Blot | |
| Biotin | |
| Armenian Hamster | |
| Apoptosis, Cell Biology, Signal Transduction | |
| PBS | |
| 23636 | |
| IgG | |
| Protein A purified |
| RL31 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
| O-linked glycoprotein-enriched fraction from rat liver nuclear envelopes raised in Armenian hamsters. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title