missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP54 Monoclonal Antibody (139CT1.1.5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537776
This item is not returnable.
View return policy
Description
NUP54 Monoclonal Antibody for Western Blot
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly repeat-containing nucleoporin subset.
Specifications
| NUP54 | |
| Monoclonal | |
| Unconjugated | |
| NUP54 | |
| 3110079L04Rik; 54 kDa nucleoporin; 54kDa; Nuclear pore complex protein Nup54; nucleoporin 54; nucleoporin 54kD; nucleoporin 54kDa; nucleoporin Nup54; Nucleoporin p54; NUP54 | |
| Mouse | |
| Protein G | |
| RUO | |
| 53371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 139CT1.1.5 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q7Z3B4 | |
| NUP54 | |
| NUP54 recombinant protein. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction