missing translation for 'onlineSavingsMsg'
Learn More

Nup53 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £470.00

Specifications

Antigen Nup53
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18491241
View Documents
Novus Biologicals
NBP1-92213-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18491221
View Documents
Novus Biologicals
NBP1-92213
0.1 mL
£470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nup53 Polyclonal antibody specifically detects Nup53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Nup53
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
35 kDa nucleoporin, Mitotic phosphoprotein 44, MP-44, MP44NUP53, NP44, Nuclear pore complex protein Nup53, nucleoporin 35kDa, Nucleoporin Nup35, nucleoporin NUP53
This antibody was developed against Recombinant Protein corresponding to amino acids: PPVRSIYDDISSPGLGSTPLTSRRQPNISVMQSPLVGVTSTPGTGQSMFSPASIGQPRKTTLSPAQLDP
Primary
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Purified
RUO
PBS (pH 7.2) and 40% Glycerol
129401
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.