missing translation for 'onlineSavingsMsg'
Learn More

NUP50 Antibody (4H7), Novus Biologicals™

Product Code. 18337389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337389 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337389 Supplier Novus Biologicals Supplier No. H00010762M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUP50 Monoclonal antibody specifically detects NUP50 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP50
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4H7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_705931
Gene Alias 50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60L, nucleoporin 50kD, nucleoporin 50kDa
Host Species Mouse
Immunogen NUP50 (NP_705931.1, 342 a.a. ~ 439 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DNEFKEKGIGTLHLKPTANQKTQLLVRADTNLGNILLNVLIPPNMPCTRTGKNNVLIVCVPNPPIDEKNATMPVTMLIRVKTSEDADELHKILLEKKD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10762
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.