missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP43 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110052

Product Code. 17147983

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DGRINLFRAD HKEAVRTIDN ADSSTLHAVT FLRTPEILTV NSIGQLKIWD FRQQGNEPSQ ILSLTGDRVP LHCVDRHPNQ QHVVATGGQD GMLSIWDVRQ GTMPVSLLKA H.

Component of the Nup107-160 subcomplex of the nuclear pore complex (NPC). The Nup107-160 subcomplex is required for the assembly of a functional NPC. The Nup107-160 subcomplex is also required for normal kinetochore microtubule attachment, mitotic progression and chromosome segregation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUP43
Polyclonal
Unconjugated
NUP43
2610016K01Rik; 2610529I12Rik; AA409950; bA350J20.1; nucleoporin 43; nucleoporin 43kDa; nucleoporin Nup43; Nup107-160 subcomplex subunit Nup43; Nup43; p42
Rabbit
Affinity Chromatography
RUO
348995, 683983
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.1 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8NFH3
NUP43
Recombinant fusion protein containing a sequence corresponding to amino acids 150-260 of human NUP43 (NP_942590.1).
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.