missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523372
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen is 89% homologous in clawed Frog.
Nucleoporin Nup35 or Mitotic phosphoprotein 44 is a nuclear protein known to be present on nuclear pore complex and is tightly associated with the nuclear membrane and lamina. This RNA associated protein belongs to the Nup53 family and has a MPPN (mitotic phosphoprotein N' end) domain. It has a small number of scattered phenylalanine-glycine repeats, but not the large domains seen in other nucleoporins. Though the exact function of NUP35 is yet to be revealed, it may function as a component of the nuclear pore complex (NPC) which are collectively referred to as nucleoporins (NUPs). It also may play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors, and may also play an important part in the association of MAD1, a transcriptional repressor with the NPC. NUP35 is widely expressed in most of the tissues.
Specifications
| NUP35 | |
| Polyclonal | |
| Unconjugated | |
| NUP35 | |
| 2310006I24Rik; 35 kDa nucleoporin; 35kDa; 5330402E05Rik; Mitotic phosphoprotein 44; Mp44; MP-44; NO44; NP44; Nuclear pore complex protein Nup53; nucleoporin 35; nucleoporin 35kDa; nucleoporin NUP35; Nucleoporin NUP53; Nup35; NUP53 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 129401, 478830, 511248, 69482 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q8NFH5, Q8R4R6 | |
| NUP35 | |
| A portion of amino acids 280-326 of human NUP35. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Canine, Monkey, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction