missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP205 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555112

Product Code. 15713041

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LDSIDFSQEI PEPLQLDFFD RAQIEQVIAN CEHKNLRGQT VCNVKLLHRV LVAEVNALQG MAAIGQRPLL MEEISTVLQY VVGRNKLLQC LHAKRHALES Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a nucleoporin, which is a subunit of the nuclear pore complex that functions in active transport of proteins, RNAs and ribonucleoprotein particles between the nucleus and cytoplasm. Mutations in this gene are associated with steroid-resistant nephrotic syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUP205
Polyclonal
Unconjugated
NUP205
205 kDa nucleoporin; 3830404O05Rik; AV248391; C7orf14; KIAA0225; mKIAA0225; NPHS13; Nuclear pore complex protein Nup205; nucleoporin 205; nucleoporin 205kDa; nucleoporin Nup205; NUP205
Rabbit
Antigen affinity chromatography
RUO
23165, 362335, 70699
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92621
NUP205
Recombinant protein corresponding to Human NUP205. Recombinant protein control fragment (Product #RP-91853).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.