missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NUP188 Polyclonal antibody specifically detects NUP188 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NUP188 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ21639, KIAA0169hNup188, nucleoporin 188kDa, nucleoporin NUP188 homolog |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human nucleoporin 188kDa using the numbering given in entry NP_056169.1 (GeneID 23511). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?