missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUP160 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848742
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848742 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848742 Supplier Invitrogen™ Supplier No. PA520565

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is A549 cell lysate. PA5-20565 can be used with blocking peptide PEP-0685.

The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-160 is part of the hetero-oligomeric complex that also contains NUP107, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP160 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP160 may be an important drug target in HIV treatment. Multiple isoforms of NUP160 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUP160
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NUP160
Gene Accession No. Q12769, Q9Z0W3
Gene Alias 160 kDa nucleoporin; 160kDa; 2810011M03Rik; AA414952; AU020188; Gene trap locus 1-13 protein; gene trap locus-13; Gtl1-13; Gtl13; GTL-13; KIAA0197; mKIAA0197; Nuclear pore complex protein Nup160; nucleoporin 160; nucleoporin 160kD; nucleoporin 160kDa; nucleoporin Nup160; NUP120; Nup160
Gene Symbols NUP160
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the carboxy terminus of human NUP160.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23279, 311182, 59015
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.