missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP155 Recombinant Rabbit Monoclonal Antibody (K01_2N66)
Description
Immunogen sequence: TAAEKKDPQG LGLHFYKHGE PEEDIVGLQA FQERLNSYKC ITDTLQELVN QSKAAPQSPS VPKKPGPPVL SSDPNMLSNE EAGHHFEQML KLSQRSKDEL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%, Pig - 100%, Cynomolgus monkey - 100%.
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
Specifications
Specifications
| Antigen | NUP155 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_2N66 |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | NUP155 |
| Gene Accession No. | O75694 |
| Gene Alias | 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?