missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP155 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119314
This item is not returnable.
View return policy
Description
NUP155 Polyclonal Antibody for Western Blot, IHC (P)
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.
Specifications
| NUP155 | |
| Polyclonal | |
| Unconjugated | |
| NUP155 | |
| 155 kDa nucleoporin; ATFB15; D930027M19Rik; KIAA0791; mKIAA0791; N155; Nuclear pore complex protein Nup155; nucleoporin 155; nucleoporin 155kD; nucleoporin 155kDa; nucleoporin Nup155; NUP155; P140 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O75694 | |
| NUP155 | |
| Recombinant protein Nucleoporin 155. The antigen corresponds to amino acid range 1154-1379 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction