missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nup153 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21248-25ul
This item is not returnable.
View return policy
Description
Nup153 Polyclonal antibody specifically detects Nup153 in Human samples. It is validated for Immunofluorescence
Specifications
| Nup153 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| CG4453, Dmel\CG4453, dmNup153, dNup153, HNUP153, N153, nuclear pore complex protein hnup153,153 kDa nucleoporin, nuclear pore complex protein Nup153, nucleoporin 153kD, nucleoporin 153kDa, Nucleoporin Nup153, Nup 153, nup153, Nup153 Nucleoporin 153 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YGVTSSTARRILQSLEKMSSPLADAKRIPSIVSSPLNSPLDRSGIDITDFQAKREKVDSQYPPVQRLMTPKPVSIATNRSVYFKPSLTPSGEFRKTNQRI | |
| 25 μg | |
| Signal Transduction | |
| 9972 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction