missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP133 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103977
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NUP133.
Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.
Specifications
| NUP133 | |
| Polyclonal | |
| Unconjugated | |
| NUP133 | |
| 133 kDa nucleoporin; hNUP133; mermaid; Nuclear pore complex protein Nup133; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin Nup133; NUP133 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 234865, 292085, 55746 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R0G9, Q8WUM0 | |
| NUP133 | |
| A synthesized peptide derived from human NUP133(Accession Q8WUM0), corresponding to amino acid residues L857-E907. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction