missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NUP133 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29044
This item is not returnable.
View return policy
Description
NUP133 Polyclonal antibody specifically detects NUP133 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NUP133 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ10814, hNUP133,133 kDa nucleoporin, MGC21133, nuclear pore complex protein Nup133, nucleoporin 133kD, nucleoporin 133kDa, Nucleoporin Nup133 | |
| between 1106 and 1156 | |
| 20 μg | |
| Signal Transduction | |
| 55746 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction