missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP107 Recombinant Rabbit Monoclonal Antibody (24GB155)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | NUP107 |
|---|---|
| Clone | 24GB155 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276040
![]() |
Invitrogen™
MA557139 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275831
|
Invitrogen™
MA557140 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUP107 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.Specifications
| NUP107 | |
| 0.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52590, P57740 | |
| 116555, 57122 | |
| A synthesized peptide derived from human Nup107 (aa 750-800). | |
| Antibody |
| 24GB155 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| NUP107 | |
| 107 kDa nucleoporin; AW541137; BcDNA.LD18761; BcDNA:LD18761; C76801; CG6743; CG6743-PA; Dmel\CG6743; Dmel_CG6743; dmNup107; NPHS11; nuclear pore complex; Nuclear pore complex protein Nup107; Nuclear pore complex protein Nup107-like protein; nucleoporin 107; Nucleoporin 107kD; nucleoporin 107kDa; nucleoporin Nup107; Nup; Nup 107; nup107; Nup107-PA; Nup170; NUP84; p105 | |
| NUP107 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title