missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP107 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519334
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.
Specifications
| NUP107 | |
| Polyclonal | |
| Unconjugated | |
| NUP107 | |
| 107 kDa nucleoporin; AW541137; BcDNA.LD18761; BcDNA:LD18761; C76801; CG6743; CG6743-PA; Dmel\CG6743; Dmel_CG6743; dmNup107; NPHS11; nuclear pore complex; Nuclear pore complex protein Nup107; Nuclear pore complex protein Nup107-like protein; nucleoporin 107; Nucleoporin 107kD; nucleoporin 107kDa; nucleoporin Nup107; Nup; Nup 107; nup107; Nup107-PA; Nup170; NUP84; p105 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 103468, 57122 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P57740, Q8BH74 | |
| NUP107 | |
| Synthetic peptide sequence (PLPAEDDNAIREH) corresponding to the internal amino acids of NUP107 (aa 735-747). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction