Learn More
Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps

Description
Allow constant airflow and minimize chance of contamination with Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Nunc sterile filter caps are made with HDPE and Thermo Scientific™ GORE™ dPTFE membrane.
- Filter membrane provides effective barrier against microorganisms
- Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
- Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
- Individually wrapped to ensure sterility
Specifications
Specifications
| Closure Color | White |
| Material | HDPE |
| For Use With (Equipment) | Nunc EasYFlasks 75 cm² |
| No. per Case | 100 |
| No. per Pack | 1 |
| Sterility | Sterile |
| Class | Class VI |
| Closure Type | Filter |
| Culture Area | 75 cm2 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.