Learn More
Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Vial Systems, manual
Quickly and easily cap and decap Thermo Scientific™ Nunc™ Cryobank tubes using these screwdrivers, available in manual and electric models to suit your application.
Brand:  Thermo Scientific™ 369644
Code : 76
Additional Details : Weight : 1.28000kg
 View more versions of this product
Product Code. 10401633
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Quickly and easily cap and decap Nunc Cryobank tubes using Thermo Scientific™ Nunc™ Screwdrivers for Cryobank and Bank-It™ Systems, available in manual and electric models to suit your application. Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.
Screwdriver socket retains the vial cap, allowing one-handed manipulation of the cap.
Specifications
Specifications
Electric Screwdriver, European Plug
2D Barcode
Red, Black
Cryobank and Bank-It Systems
Manual Screwdriver
Cryobank, Bank-It
European