Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
Descrizione
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Features:
- Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
- One-piece seamless design reduces possibility of leaking through a seam.
- Shallow and deep indents allow for excellent manual and automated handling.
- Proprietary manufacturing process allows for greater impact resistance
- Vertical expanded surfaces version facilitates emptying and reduces retention of product
Recommended for:
- Cell culture expansion
- Vaccine production
- Antibody production
Specifica
Specifica
| Bottle Surface | Smooth, no indent |
| Color | Clear |
| For Use With (Application) | Cell Culture Expansion, Vaccine Production, Antibody Production |
| Material | Polystyrene |
| No. per Case | 20 |
| No. per Pack | 20 |
| Packaging | 20/Pk., 20/Cs. |
| Sterility | Sterile |
| Cell Growth Area | 850 cm2 |
| Closure Included | Yes |
| Vedi altri risultati |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.