Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles
Product Code. 11727634
Change view
Click to view available options
Cell Growth Area:
1450 cm2
850 cm2
Bottle Surface:
Pleated, no indent
Smooth, deep indent
Smooth, no indent
Smooth, shallow indent
Closure Type:
EZ Twist Non-vented Cap
EZ Twist Vented Cap
Easy on/off, Non-vented Cap
Easy on/off, Vented Cap
Packaging:
2/Pk., 20/Cs.
2/Pk., 20/Cs. (Double-bagged)
20/Pk., 20/Cs.
20/Pk., 20/Cs. (Double-bagged)
Unit Size:
Pack of 20
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cell Growth Area Bottle Surface Closure Type Packaging Unit Size
11727634 1450 cm2 Pleated, no indent Easy on/off, Vented Cap 20/Pk., 20/Cs. (Double-bagged) Pack of 20
11947894 850 cm2 Smooth, no indent Easy on/off, Vented Cap 2/Pk., 20/Cs. Pack of 20
11967894 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs. Pack of 20
11977894 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs. (Double-bagged) Pack of 20
11957894 850 cm2 Smooth, no indent Easy on/off, Non-vented Cap 2/Pk., 20/Cs. Pack of 20
11987894 850 cm2 Smooth, shallow indent EZ Twist Vented Cap 2/Pk., 20/Cs. Pack of 20
11997894 850 cm2 Smooth, deep indent EZ Twist Vented Cap 2/Pk., 20/Cs. Pack of 20
11907904 850 cm2 Smooth, shallow indent EZ Twist Non-vented Cap 2/Pk., 20/Cs. Pack of 20
11917904 850 cm2 Smooth, shallow indent EZ Twist Non-vented Cap 20/Pk., 20/Cs. (Double-bagged) Pack of 20
12311393 850 cm2 Smooth, deep indent EZ Twist Non-vented Cap 2/Pk., 20/Cs. (Double-bagged) Pack of 20
12311643 850 cm2 Smooth, deep indent EZ Twist Non-vented Cap 20/Pk., 20/Cs. Pack of 20
11947904 850 cm2 Smooth, deep indent EZ Twist Non-vented Cap 20/Pk., 20/Cs. (Double-bagged) Pack of 20
11937894 1450 cm2 Pleated, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs. (Double-bagged) Pack of 20
11927894 1450 cm2 Pleated, no indent Easy on/off, Non-vented Cap 20/Pk., 20/Cs. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 11727634 Supplier Thermo Scientific™ Supplier No. 141744
Estimated Shipment: 28-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Features:
  • Nunc PS Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6
  • One-piece seamless design reduces possibility of leaking through a seam.
  • Shallow and deep indents allow for excellent manual and automated handling.
  • Proprietary manufacturing process allows for greater impact resistance
  • Vertical expanded surfaces version facilitates emptying and reduces retention of product
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Pleated, no indent
Material Polystyrene
No. per Case 20
No. per Pack 20
Packaging 20/Pk., 20/Cs. (Double-bagged)
Cell Growth Area 1450 cm2
Closure Included Yes
Closure Type Easy on/off, Vented Cap
Product Type Roller Bottle
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.