missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ NucleoLink™ / TopYield™ Frame

Product Code. 11331575
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11331575 Red Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11331575 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 249182

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for liquid phase PCR, immuno-PCR and hybridization assays, the unique design of these frames offer error-free flexibility, accurate reading and efficient washing.

Ideal for liquid phase PCR, immuno-PCR and hybridization assays due to the heat resistant resin, the unique design of Thermo ScientificTM NuncTM NucleoLinkTM Frames and breakable modules offer error-free flexibility; each well locks in the frame and maintains the same height to provide accurate reading and efficient washing.

Push the limits of your analysis and evaluate your performance with frames and strips that are ideal for binding nucleotides (e.g., DNA and RNA) and for hybridization assays.
  • Use in place of time-consuming methods such as gel electrophoresis and Southern blotting
  • Usable with any “ELISA-like” detection and measurement method
  • Compatible with standard microplate readers
  • Consists of 1 × 8 strips fitted into a 96-well format frame
  • V-shaped wells with flat, readable bottoms and thin walls (0.35mm)
  • Withstand temperatures from -20° to +120°C
  • Working volume: Strips 100μL
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Description Nunc NucleoLink/TopYield Frame
Array 8 x 12
Packaging 72 Cases
Color Red
Product Type Frame
Material ABS Plastic
No. of Wells 96
Volume (Metric) Working 100 μL
Well Shape Conical Bottom (V-well)
For Use With (Application) Liquid phase PCR or Immuno-PCR
Max. Heat Stability 90°C
No. per Case 72
No. per Pack 6
Plate Compatibility 96-Well Plates
Quantity 72
Length (Metric) 128 mm
Width (Metric) 86 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.