missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ MicroWell™ MiniTrays
BIOPHARMA

Product Code. 10112791
Change view
Click to view available options
Well Count:
60
72
Closure Included:
No
Yes
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 400
Pack of 480
Product Code. Well Count Closure Included Sterility unitSize
10112791 72 Yes Non-sterile Pack of 480
10192831 72 Yes Non-sterile Pack of 100
10104371 60 Yes Non-sterile Pack of 100
10204221 60 Yes Sterile Pack of 480
10326141 60 No Sterile Pack of 400
5 options
This item is not returnable. View return policy

Product Code. 10112791

Brand: Thermo Scientific™ 470378

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.

Create more space in lab freezers and inubators with Thermo Scientific™ Nunc™ MicroWell™ MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.

  • Limited adsorption of proteins
  • Lids have keyed tabs for proper orientation
  • Large raised alphanumeric coordinates
  • Unique surface treatment to maximize sample and reagent mixing
  • MiniTrays for serological applications have slightly smaller well dimensions
TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Nunclon Delta
Type Cell Culture Dish
Well Count 72
Includes Lid
No. per Case 480
No. per Pack 120
Length (Metric) 84 mm
Width (Metric) 59 mm
Height (Metric) 59 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.