missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nunc MaxiSorp™ flat-bottom

Product Code. 16567010
Click to view available options
Color:
Clear
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16567010

Brand: Invitrogen™ 44240421

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High protein-binding capacity polystyrene 96 well ELISA plates that are recommended for use with eBioscience Ready-SET-Go! ELISA reagent sets, as well as other matched antibody pairs for ELISA.

Low-protein binding capacity plates yield low signal or inconsistent data. Use of high protein-binding capacity polystyrene 96 well ELISA plates is essential for optimal assay performance.
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Product Line Nunc MaxiSorp
Color Clear
Product Type ELISA Microplate
No. of Wells 96
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 96 tests

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.