missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nunc MaxiSorp™ flat-bottom

Product Code. 11530627
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11530627 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11530627 Supplier Invitrogen™ Supplier No. 44240421

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High protein-binding capacity polystyrene 96 well ELISA plates that are recommended for use with eBioscience Ready-SET-Go! ELISA reagent sets, as well as other matched antibody pairs for ELISA.

Low-protein binding capacity plates yield low signal or inconsistent data. Use of high protein-binding capacity polystyrene 96 well ELISA plates is essential for optimal assay performance.
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Colorimetric
Product Line Nunc MaxiSorp
Color Clear
Product Type ELISA Microplate
No. of Wells 96
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 96 tests

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.