missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ EasYDish™ Dishes with Nunclon™ Supra Surface

Product Code. 30408793
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Culture Area:
145 cm2
21.5 cm2
56.7 cm2
8.8 cm2
Height (Metric):
13 mm
16 mm
17 mm
21 mm
Unit Size:
Pack of 15
Pack of 150
Pack of 24
Pack of 240
Pack of 28
Pack of 280
Pack of 50
Pack of 500
Pack of 8
Pack of 80
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Culture Area Height (Metric) Unit Size
30408793 35 mm 8.8 cm2 13 mm Pack of 500
17947422 35 mm 8.8 cm2 13 mm Pack of 50
30408804 60 mm 21.5 cm2 16 mm Pack of 280
17987422 60 mm 21.5 cm2 16 mm Pack of 28
30408787 100 mm 56.7 cm2 21 mm Pack of 240
17977422 100 mm 56.7 cm2 21 mm Pack of 24
17907432 100 mm 56.7 cm2 17 mm Pack of 15
30408794 100 mm 56.7 cm2 17 mm Pack of 150
30408791 150 mm 145 cm2 21 mm Pack of 80
17967422 150 mm 145 cm2 21 mm Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 30408793 Supplier Thermo Scientific™ Supplier No. 150470
  • ONLINE EXCLUSIVE PRICE
    £351.00
    £329.91 / Pack of 500
    Save £21.09
    6% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc EasYDish dishes with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The beveled grip ring is easy to hold and stays secure in your gloved hand. Available in four sizes: 35, 60, 100, and 150 mm. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc EasYDish Dishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Culture Area 8.8 cm2
Disposable Single-use
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Nunclon Supra
Type Cell Culture Dish
No. per Case 500 Dishes
No. per Pack 10 Dishes
Height (Metric) 13 mm
Diameter (Metric) 35 mm
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.