Learn More
Thermo Scientific™ Nunc™ 96-Well, Nunclon Delta-Treated, U-Shaped-Bottom Microplate
BIOPHARMA
Product Code. 10337382
Change view
Click to view available options
Unit Size:
Pack of 160
Product Code. unitSize
10337382 Pack of 160
1 options
Product Code. 10337382 Supplier Thermo Scientific™ Supplier No. 168136
Only null left
Add to basket
Add to basket
Color:Clear; Sterility:Sterile; Lid:With Lid
Thermo Scientific™ Nunc 96-Well Polystyrene Round Bottom Microwell Plates are optimized for robotics, automated readers and liquid handling systems. 96-well round bottom (U) microplates are ideal for agglutination assays.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Certified Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Lid With Lid
Array 8 x 12
Packaging 10 Pk., 160 Case
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 300 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Round Bottom (U-well)
No. per Case 160
No. per Pack 10
Vis mere Vis mindre
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.