missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUMB Recombinant Rabbit Monoclonal Antibody (23GB1070), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | NUMB |
|---|---|
| Clone | 23GB1070 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088470
|
Invitrogen™
MA554048 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088159
|
Invitrogen™
MA554049 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUMB Recombinant Monoclonal antibody specifically detects NUMB in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| NUMB | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P49757, Q2LC84, Q9QZS3 | |
| 18222, 29419, 8650 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1070 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171 | |
| A synthetic peptide of human NUMB (413-651AA). | |
| Primary | |
| -20°C | |
| Numb |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title