missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUMB Polyclonal Antibody

Product Code. 11525403
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11525403 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11525403 Supplier Invitrogen™ Supplier No. PA116831

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The target sequence has 90% sequence homology with rat.

Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUMB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene Numb
Gene Accession No. P49757, Q9QZS3
Gene Alias c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171
Gene Symbols Numb
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of mouse NUMB (between residues 600-653).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18222, 395639, 8650
Target Species Human, Mouse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.