missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUMB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119316
This item is not returnable.
View return policy
Description
NUMB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.
Specifications
| NUMB | |
| Polyclonal | |
| Unconjugated | |
| Numb | |
| c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 18222, 8650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P49757, Q9QZS3 | |
| Numb | |
| Recombinant protein Numb Homolog. The antigen corresponds to amino acid range 376-482 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction